missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab1A Mouse anti-Human, Mouse, Clone: 4G10, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP3-11043-100UG
This item is not returnable.
View return policy
Description
Rab1A Monoclonal specifically detects Rab1A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Rab1A | |
Monoclonal | |
1 mg/ml | |
Western Blot 1:1000, Immunoprecipitation | |
DKFZp564B163, member RAS oncogene family, RAB1A, member RAS oncogene family, ras-related protein Rab-1A | |
Full length recombinant Rab1A protein (NP_004152.1) | |
100 μg | |
Primary | |
Human, Mouse | |
Purified |
Western Blot, Immunoprecipitation | |
4G10 | |
Unconjugated | |
PBS (pH 7.4), 50% Glycerol | |
Mouse | |
Protein G purified | |
RUO | |
5861 | |
Store at -20°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction