missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rab1A Mouse anti-Human, Mouse, Clone: 4G10, Novus Biologicals™
Mouse Monoclonal Antibody
2450.00 SEK - 5640.00 SEK
Specifications
Antigen | Rab1A |
---|---|
Clone | 4G10 |
Concentration | 1 mg/ml |
Dilution | Western Blot 1:1000, Immunoprecipitation |
Applications | Western Blot, Immunoprecipitation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18337631
|
Bio-Techne
NBP3-11043-25UG |
25 μg |
2450.00 SEK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18316202
|
Bio-Techne
NBP3-11043-100UG |
100 μg |
5640.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rab1A Monoclonal specifically detects Rab1A in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Rab1A | |
1 mg/ml | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Mouse | |
DKFZp564B163, member RAS oncogene family, RAB1A, member RAS oncogene family, ras-related protein Rab-1A | |
Full length recombinant Rab1A protein (NP_004152.1) | |
Primary | |
Store at -20°C. |
4G10 | |
Western Blot 1:1000, Immunoprecipitation | |
Monoclonal | |
Purified | |
RUO | |
PBS (pH 7.4), 50% Glycerol | |
5861 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title