missing translation for 'onlineSavingsMsg'
Learn More

Rabies Virus Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ Rabies Virus Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia MS-549-P

 View more versions of this product

Product Code. 12601617

This item is not returnable. View return policy

Description

Description

Host Species: Mouse

Clone: Rab-50

Isotype: IgG2b/κ

Species Reactivity: Rabies virus of strains SAD-Vnukovo and Pitman Moore

Epitope: Not determined

Immunogen: Rabies Virus of strains SAD-Vnukovo and Pitman Moore Rab-50 antibody reacts with viral glycoprotein of rabies virus strains SAD-Vnukovo and Pitman-Moore. It can be used for detecting rabies virus by ELISA and Western. It is capable of neutralizing rabies virus.

Recommended for:

  • ELISA
  • Neutralization in vitro

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rabies Virus Ab-1
Rab-50
Unconjugated
Rabies Virus of strains SAD-Vnukovo and Pitman Moore
RUO
Primary
Concentrated
Monoclonal
200μg/mL
Mouse
1 mL
Infectious Disease
Rabies virus
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Rabies Virus Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™ > 1mL; 200μg/mL; Unlabeled; Purified with BSA and azide

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.