Learn More
Rabies Virus Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™
Provide accurate, reproducible results with the Epredia™ Rabies Virus Ab-1, Mouse Monoclonal Antibody.
Brand: Epredia MS-549-P1
Description
Host Species: Mouse
Clone: Rab-50
Isotype: IgG2b/κ
Species Reactivity: Rabies virus of strains SAD-Vnukovo and Pitman Moore
Epitope: Not determined
Immunogen: Rabies Virus of strains SAD-Vnukovo and Pitman Moore Rab-50 antibody reacts with viral glycoprotein of rabies virus strains SAD-Vnukovo and Pitman-Moore. It can be used for detecting rabies virus by ELISA and Western. It is capable of neutralizing rabies virus.
Recommended for:
- ELISA
- Neutralization in vitro
Note:
USA: RUO; Int'l: RUO

Specifications
Rabies Virus Ab-1 | |
Rab-50 | |
Unconjugated | |
Rabies Virus of strains SAD-Vnukovo and Pitman Moore | |
RUO | |
Primary | |
Concentrated |
Monoclonal | |
200μg/mL | |
Mouse | |
500 μL | |
Infectious Disease | |
Rabies virus | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.