missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine IL10 / Interleukin-10 Protein

A DNA sequence encoding the Bos taurus (Bovine) IL10 / Interleukin-10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9204-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16017841

  • 12410.00 SEK / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_776513.1
IL10 / Interleukin-10 Protein
Research Use Only
IL10
Recombinant Protein
Yeast
SRDASTLSDS SCIHLPTSLP HMLRELRAAF GKVKTFFQMK DQLHSLLLTQ SLLDDFKGYL GCQALSEMIQ FYLEEVMPQA ENHGPDIKEH VNSLGEKLKT LRLRLRRCHR FLPCENKSKA VEKVKRVFSE LQERGVYKAM SEFDIFINYI ETYMTTKMQK
Greater than 90% as determined by SDS-PAGE.
281246
100 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers