missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine IL10 / Interleukin-10 Protein
A DNA sequence encoding the Bos taurus (Bovine) IL10 / Interleukin-10, was expressed in the hosts and tags indicated.
3490.00 SEK - 31480.00 SEK
Specifications
Accession Number | NP_776513.1 |
---|---|
Gene ID (Entrez) | 281246 |
Name | IL10 / Interleukin-10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16027841
|
enQuireBio™
QP9204-ye-10ug |
10 μg | |||||||
16067841
|
enQuireBio™
QP9204-ye-50ug |
50 μg | |||||||
16017841
|
enQuireBio™
QP9204-ye-100ug |
100 μg |
12410.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16047841
|
enQuireBio™
QP9204-ye-200ug |
200 μg | |||||||
16057841
|
enQuireBio™
QP9204-ye-500ug |
500 μg |
29995.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16037841
|
enQuireBio™
QP9204-ye-1mg |
1 mg | |||||||
Specifications
NP_776513.1 | |
IL10 / Interleukin-10 Protein | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
281246 | |
Research Use Only | |
IL10 | |
Recombinant Protein | |
Yeast | |
SRDASTLSDS SCIHLPTSLP HMLRELRAAF GKVKTFFQMK DQLHSLLLTQ SLLDDFKGYL GCQALSEMIQ FYLEEVMPQA ENHGPDIKEH VNSLGEKLKT LRLRLRRCHR FLPCENKSKA VEKVKRVFSE LQERGVYKAM SEFDIFINYI ETYMTTKMQK | |
Greater than 90% as determined by SDS-PAGE. |