missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Metalloproteinase inhibitor 4 Protein

A DNA sequence encoding the Bos taurus (Bovine) Metalloproteinase inhibitor 4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6796-ye-500ug

 View more versions of this product

Product Code. 16055920

  • 31245.00 SEK / 500µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001039336.1
Metalloproteinase inhibitor 4 Protein
Research Use Only
TIMP4
Recombinant Protein
Yeast
CSCAPAHPQQ HVCHSALAIR AKISSEKVVP ASTDPADPQK MIRYEIKQIK MFKGFEKVND IQYIYTPFDS SLCGVKLEAN SQKRYLLTGQ ILSDGKVFVH LCNYIEPWEN LSFLQRESLN HHYHLNCGCQ ITTCYAVPCT ISAPNECLWT DWLLERKLYG YQAQHYVCMK HVDGSCSWYQ GRLPLRKEFV DIIQP
Greater than 90% as determined by SDS-PAGE.
317694
500 μg
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers