missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Metalloproteinase inhibitor 4 Protein

A DNA sequence encoding the Bos taurus (Bovine) Metalloproteinase inhibitor 4, was expressed in the hosts and tags indicated.

3615.00 SEK - 32610.00 SEK

Specifications

Accession Number NP_001039336.1
Gene ID (Entrez) 317694
Name Metalloproteinase inhibitor 4 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16025920
View Documents Promotion Details
enQuireBio™
QP6796-ye-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16065920
View Documents Promotion Details
enQuireBio™
QP6796-ye-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16015920
View Documents Promotion Details
enQuireBio™
QP6796-ye-100ug
100 μg
12935.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16045920
View Documents Promotion Details
enQuireBio™
QP6796-ye-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16055920
View Documents Promotion Details
enQuireBio™
QP6796-ye-500ug
500 μg
31245.00 SEK
500µg
Please to purchase this item. Need a web account? Register with us today!
16035920
View Documents Promotion Details
enQuireBio™
QP6796-ye-1mg
1 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Specifications

NP_001039336.1
Metalloproteinase inhibitor 4 Protein
Not determined.
Full Length
Bovine
His
Tris-based buffer, 50% glycerol
317694
Research Use Only
TIMP4
Recombinant Protein
Yeast
CSCAPAHPQQ HVCHSALAIR AKISSEKVVP ASTDPADPQK MIRYEIKQIK MFKGFEKVND IQYIYTPFDS SLCGVKLEAN SQKRYLLTGQ ILSDGKVFVH LCNYIEPWEN LSFLQRESLN HHYHLNCGCQ ITTCYAVPCT ISAPNECLWT DWLLERKLYG YQAQHYVCMK HVDGSCSWYQ GRLPLRKEFV DIIQP
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications