missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog IL-18 / IL-1F4 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) IL-18 / IL-1F4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8263-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001003169.1 | |
IL-18 / IL-1F4 Protein | |
Research Use Only | |
IL18 | |
Recombinant Protein | |
E. coli | |
YFGKLEPKLS IIRNLNDQVL FVNEGNQPVF EDMPDSDCTD NAPHTIFIIY MYKDSLTRGL AVTISVKYKT MSTLSCKNKT ISFQKMSPPD SINDEGNDII FFQRSVPGHD DKIQFESSLY KGHFLACKKE NDLFKLILKD KDENGDKSIM FTVQNKS | |
Greater than 90% as determined by SDS-PAGE. |
403796 | |
500 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |