missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog IL-18 / IL-1F4 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) IL-18 / IL-1F4, was expressed in the hosts and tags indicated.
3070.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_001003169.1 |
---|---|
Gene ID (Entrez) | 403796 |
Name | IL-18 / IL-1F4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093621
|
enQuireBio™
QP8263-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013631
|
enQuireBio™
QP8263-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003631
|
enQuireBio™
QP8263-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023631
|
enQuireBio™
QP8263-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033631
|
enQuireBio™
QP8263-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043631
|
enQuireBio™
QP8263-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001003169.1 | |
IL-18 / IL-1F4 Protein | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |
403796 | |
Research Use Only | |
IL18 | |
Recombinant Protein | |
E. coli | |
YFGKLEPKLS IIRNLNDQVL FVNEGNQPVF EDMPDSDCTD NAPHTIFIIY MYKDSLTRGL AVTISVKYKT MSTLSCKNKT ISFQKMSPPD SINDEGNDII FFQRSVPGHD DKIQFESSLY KGHFLACKKE NDLFKLILKD KDENGDKSIM FTVQNKS | |
Greater than 90% as determined by SDS-PAGE. |