missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli DNA-binding protein HU-alpha Protein
A DNA sequence encoding the Escherichia coli O157:H7 DNA-binding protein HU-alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7329-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_312950.1 | |
DNA-binding protein HU-alpha Protein | |
Research Use Only | |
hupA | |
Recombinant Protein | |
E. coli | |
MNKTQLIDVI AEKAELSKTQ AKAALESTLA AITESLKEGD AVQLVGFGTF KVNHRAERTG RNPQTGKEIK IAAANVPAFV SGKALKDAVK | |
Greater than 94.2% as determined by SDS-PAGE. |
914933 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction