missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli DNA-binding protein HU-alpha Protein

A DNA sequence encoding the Escherichia coli O157:H7 DNA-binding protein HU-alpha, was expressed in the hosts and tags indicated.

3070.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_312950.1
Gene ID (Entrez) 914933
Name DNA-binding protein HU-alpha Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16038240
View Documents
enQuireBio™
QP7329-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16078240
View Documents
enQuireBio™
QP7329-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16028240
View Documents
enQuireBio™
QP7329-ec-100ug
100 μg
11590.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16058240
View Documents
enQuireBio™
QP7329-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16068240
View Documents
enQuireBio™
QP7329-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16048240
View Documents
enQuireBio™
QP7329-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_312950.1
DNA-binding protein HU-alpha Protein
Not determined.
Full Length
E. coli
Tris-based buffer, 50% glycerol
914933
Research Use Only
hupA
Recombinant Protein
MNKTQLIDVI AEKAELSKTQ AKAALESTLA AITESLKEGD AVQLVGFGTF KVNHRAERTG RNPQTGKEIK IAAANVPAFV SGKALKDAVK
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.