missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli LptC Protein

A DNA sequence encoding the Escherichia coli (strain K12) LptC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7332-ye-200ug

 View more versions of this product

Product Code. 16058270

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

WP_000030537.1
200 μg
Not determined.
Full Length
E. coli
His
Tris-based buffer, 50% glycerol
LptC Protein
Research Use Only
lptC
Recombinant Protein
Yeast
MSKARRWVII VLSLAVLVMI GINMAEKDDT AQVVVNNNDP TYKSEHTDTL VYNPEGALSY RLIAQHVEYY SDQAVSWFTQ PVLTTFDKDK IPTWSVKADK AKLTNDRMLY LYGHVEVNAL VPDSQLRRIT TDNAQINLVT QDVTSEDLVT LYGTTFNSSG LKMRGNLRSK NAELIEKVRT SYEIQNKQTQ P
Greater than 90% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers