missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli LptC Protein
A DNA sequence encoding the Escherichia coli (strain K12) LptC, was expressed in the hosts and tags indicated.
3615.00 SEK - 31480.00 SEK
Specifications
Accession Number | WP_000030537.1 |
---|---|
Name | LptC Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | lptC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16038270
|
enQuireBio™
QP7332-ye-10ug |
10 μg | |||||||
16078270
|
enQuireBio™
QP7332-ye-50ug |
50 μg | |||||||
16028270
|
enQuireBio™
QP7332-ye-100ug |
100 μg |
12410.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16058270
|
enQuireBio™
QP7332-ye-200ug |
200 μg | |||||||
16068270
|
enQuireBio™
QP7332-ye-500ug |
500 μg |
29995.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16048270
|
enQuireBio™
QP7332-ye-1mg |
1 mg | |||||||
Specifications
WP_000030537.1 | |
Research Use Only | |
lptC | |
Recombinant Protein | |
Yeast | |
MSKARRWVII VLSLAVLVMI GINMAEKDDT AQVVVNNNDP TYKSEHTDTL VYNPEGALSY RLIAQHVEYY SDQAVSWFTQ PVLTTFDKDK IPTWSVKADK AKLTNDRMLY LYGHVEVNAL VPDSQLRRIT TDNAQINLVT QDVTSEDLVT LYGTTFNSSG LKMRGNLRSK NAELIEKVRT SYEIQNKQTQ P | |
Greater than 90% as determined by SDS-PAGE. |
LptC Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |