missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli LptC Protein
A DNA sequence encoding the Escherichia coli (strain K12) LptC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7332-ye-50ug
This item is not returnable.
View return policy
Specifications
WP_000030537.1 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
LptC Protein | |
Research Use Only | |
lptC | |
Recombinant Protein | |
Yeast | |
MSKARRWVII VLSLAVLVMI GINMAEKDDT AQVVVNNNDP TYKSEHTDTL VYNPEGALSY RLIAQHVEYY SDQAVSWFTQ PVLTTFDKDK IPTWSVKADK AKLTNDRMLY LYGHVEVNAL VPDSQLRRIT TDNAQINLVT QDVTSEDLVT LYGTTFNSSG LKMRGNLRSK NAELIEKVRT SYEIQNKQTQ P | |
Greater than 90% as determined by SDS-PAGE. |