missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DDB1- and CUL4-associated factor 7 Protein
A DNA sequence encoding the Homo sapiens (Human) DDB1- and CUL4-associated factor 7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6885-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_005819.3 | |
DDB1- and CUL4-associated factor 7 Protein | |
Research Use Only | |
DCAF7 | |
Recombinant Protein | |
E. coli | |
MSLHGKRKEI YKYEAPWTVY AMNWSVRPDK RFRLALGSFV EEYNNKVQLV GLDEESSEFI CRNTFDHPYP TTKLMWIPDT KGVYPDLLAT SGDYLRVWRV GETETRLECL LNNNKNSDFC APLTSFDWNE VDPYLLGTSS IDTTCTIWGL ETGQVLGRVN LVSGHVKTQL IAHDKEVYDI AFSRAGGGRD MFASVGADGS VRMFDLRHLE HSTIIYEDPQ HHPLLRLCWN KQDPNYLATM AMDGMEVVIL DVRVPCTPVA RLNNHRACVN GIAWAPHSSC HICTAADDHQ ALIWDIQQMP RAIEDPILAY TAEGEINNVQ WASTQPDWIA ICYNNCLEIL RV | |
Greater than 90% as determined by SDS-PAGE. |
10238 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |