missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Galactose-1-phosphate uridylyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Galactose-1-phosphate uridylyltransferase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6067-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_000146.2 | |
Galactose-1-phosphate uridylyltransferase Protein | |
Research Use Only | |
GALT | |
Recombinant Protein | |
E. coli | |
MSRSGTDPQQ RQQASEADAA AATFRANDHQ HIRYNPLQDE WVLVSAHRMK RPWQGQVEPQ LLKTVPRHDP LNPLCPGAIR ANGEVNPQYD STFLFDNDFP ALQPDAPSPG PSDHPLFQAK SARGVCKVMC FHPWSDVTLP LMSVPEIRAV VDAWASVTEE LGAQYPWVQI FENKGAMMGC SNPHPHCQVW ASSFLPDIAQ REERSQQAYK SQHGEPLLME YSRQELLRKE RLVLTSEHWL VLVPFWATWP YQTLLLPRRH VRRLPELTPA ERDDLASIMK KLLTKYDNLF ETSFPYSMGW HGAPTGSEAG ANWNHWQLHA HYYPPLLRSA TVRKFMVGYE MLAQAQRDLT PEQAAERLRA LPEVHYHLGQ KDRETATIA | |
Greater than 90% as determined by SDS-PAGE. |
2592 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction