missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Galactose-1-phosphate uridylyltransferase Protein
A DNA sequence encoding the Homo sapiens (Human) Galactose-1-phosphate uridylyltransferase, was expressed in the hosts and tags indicated.
1640.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_000146.2 |
---|---|
Gene ID (Entrez) | 2592 |
Name | Galactose-1-phosphate uridylyltransferase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15979739
|
enQuireBio™
QP6067-ec-10ug |
10 μg | N/A | ||||||
15919749
|
enQuireBio™
QP6067-ec-50ug |
50 μg | N/A | ||||||
15969739
|
enQuireBio™
QP6067-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15999739
|
enQuireBio™
QP6067-ec-200ug |
200 μg | N/A | ||||||
15909749
|
enQuireBio™
QP6067-ec-500ug |
500 μg | N/A | ||||||
15989739
|
enQuireBio™
QP6067-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000146.2 | |
Galactose-1-phosphate uridylyltransferase Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2592 | |
Research Use Only | |
GALT | |
Recombinant Protein | |
E. coli | |
MSRSGTDPQQ RQQASEADAA AATFRANDHQ HIRYNPLQDE WVLVSAHRMK RPWQGQVEPQ LLKTVPRHDP LNPLCPGAIR ANGEVNPQYD STFLFDNDFP ALQPDAPSPG PSDHPLFQAK SARGVCKVMC FHPWSDVTLP LMSVPEIRAV VDAWASVTEE LGAQYPWVQI FENKGAMMGC SNPHPHCQVW ASSFLPDIAQ REERSQQAYK SQHGEPLLME YSRQELLRKE RLVLTSEHWL VLVPFWATWP YQTLLLPRRH VRRLPELTPA ERDDLASIMK KLLTKYDNLF ETSFPYSMGW HGAPTGSEAG ANWNHWQLHA HYYPPLLRSA TVRKFMVGYE MLAQAQRDLT PEQAAERLRA LPEVHYHLGQ KDRETATIA | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title