missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human GTPase ERas Protein

A DNA sequence encoding the Homo sapiens (Human) GTPase ERas, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8776-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16006421

  • 5980.00 SEK / 100µg
Estimated Shipment: 05-09-2024
to see stock.

Description

Description

Specifications

Specifications

NP_853510.1
GTPase ERas Protein
Research Use Only
ERAS
Recombinant Protein
E. coli
LPTKPGTFDL GLATWSPSFQ GETHRAQARR RDVGRQLPEY KAVVVGASGV GKSALTIQLN HQCFVEDHDP TIQDSYWKEL TLDSGDCILN VLDTAGQAIH RALRDQCLAV CDGVLGVFAL DDPSSLIQLQ QIWATWGPHP AQPLVLVGNK CDLVTTAGDA HAAAAALAHS WGAHFVETSA KTRQGVEEAF SLLVHEIQRV QEAMAKEPMA RSCREKTRHQ KATCHCGC
Greater than 90% as determined by SDS-PAGE.
3266
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers