missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GTPase ERas Protein
A DNA sequence encoding the Homo sapiens (Human) GTPase ERas, was expressed in the hosts and tags indicated.
1580.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_853510.1 |
---|---|
Gene ID (Entrez) | 3266 |
Name | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016421
|
enQuireBio™
QP8776-ec-10ug |
10 μg | N/A | ||||||
16056421
|
enQuireBio™
QP8776-ec-50ug |
50 μg | N/A | ||||||
16006421
|
enQuireBio™
QP8776-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16036421
|
enQuireBio™
QP8776-ec-200ug |
200 μg | N/A | ||||||
16046421
|
enQuireBio™
QP8776-ec-500ug |
500 μg | N/A | ||||||
16026421
|
enQuireBio™
QP8776-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_853510.1 | |
GTPase ERas Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
Tris-based buffer, 50% glycerol |
3266 | |
Research Use Only | |
ERAS | |
Recombinant Protein | |
LPTKPGTFDL GLATWSPSFQ GETHRAQARR RDVGRQLPEY KAVVVGASGV GKSALTIQLN HQCFVEDHDP TIQDSYWKEL TLDSGDCILN VLDTAGQAIH RALRDQCLAV CDGVLGVFAL DDPSSLIQLQ QIWATWGPHP AQPLVLVGNK CDLVTTAGDA HAAAAALAHS WGAHFVETSA KTRQGVEEAF SLLVHEIQRV QEAMAKEPMA RSCREKTRHQ KATCHCGC |