missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-DPB1 Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-DPB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8814-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_002112.3 | |
HLA-DPB1 Protein | |
Research Use Only | |
HLA-DPB1 | |
Recombinant Protein | |
E. coli | |
RATPENYLFQ GRQECYAFNG TQRFLERYIY NREEFARFDS DVGEFRAVTE LGRPAAEYWN SQKDILEEKR AVPDRMCRHN YELGGPMTLQ RRVQPRVNVS PSKKGPLQHH NLLVCHVTDF YPGSIQVRWF LNGQEETAGV VSTNLIRNGD WTFQILVMLE MTPQQGDVYT CQVEHTSLDS PVTVEWKAQS DSARSK | |
Greater than 90% as determined by SDS-PAGE. |
3115 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction