missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HLA-DPB1 Protein
A DNA sequence encoding the Homo sapiens (Human) HLA-DPB1, was expressed in the hosts and tags indicated.
1580.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_002112.3 |
---|---|
Gene ID (Entrez) | 3115 |
Name | HLA-DPB1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16016631
|
enQuireBio™
QP8814-ec-10ug |
10 μg | N/A | ||||||
16056631
|
enQuireBio™
QP8814-ec-50ug |
50 μg | N/A | ||||||
16006631
|
enQuireBio™
QP8814-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16036631
|
enQuireBio™
QP8814-ec-200ug |
200 μg | N/A | ||||||
16046631
|
enQuireBio™
QP8814-ec-500ug |
500 μg | N/A | ||||||
16026631
|
enQuireBio™
QP8814-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_002112.3 | |
HLA-DPB1 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3115 | |
Research Use Only | |
HLA-DPB1 | |
Recombinant Protein | |
E. coli | |
RATPENYLFQ GRQECYAFNG TQRFLERYIY NREEFARFDS DVGEFRAVTE LGRPAAEYWN SQKDILEEKR AVPDRMCRHN YELGGPMTLQ RRVQPRVNVS PSKKGPLQHH NLLVCHVTDF YPGSIQVRWF LNGQEETAGV VSTNLIRNGD WTFQILVMLE MTPQQGDVYT CQVEHTSLDS PVTVEWKAQS DSARSK | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title