missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-31 / IL31 Protein
A DNA sequence encoding the Human IL-31 / IL31, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10328-ec-250ug
This item is not returnable.
View return policy
Specifications
NP_001014358.1 | |
IL-31 / IL31 Protein | |
Research Use Only | |
IL31 | |
The specific activity is determined by inducing STAT3 activation using human U-87 MG cells. 5 ng/mL of rHuIL-31 can effectively induce STAT3 activation. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH7.4, lyophilized |
386653 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
SHTLPVRLLR PSDDVQKIVE ELQSLSKMLL KDVEEEKGVL VSQNYTLPCL SPDAQPPNNI HSPAIRAYLK TIRQLDNKSV IDEIIEHLDK LIFQDAPETN ISVPTDTHEC KRFILTISQQ FSECMDLALK SLTSGAQQAT T | |
Greater than 97% as determined by SDS-PAGE and HPLC. |