missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-31 / IL31 Protein

A DNA sequence encoding the Human IL-31 / IL31, was expressed in the hosts and tags indicated.

1080.00 SEK - 33925.00 SEK

Specifications

Accession Number NP_001014358.1
Gene ID (Entrez) 386653
Name IL-31 / IL31 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15963518
View Documents Promotion Details
enQuireBio™
QP10328-ec-2ug
2 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15943518
View Documents Promotion Details
enQuireBio™
QP10328-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15933518
View Documents Promotion Details
enQuireBio™
QP10328-ec-100ug
100 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15953518
View Documents Promotion Details
enQuireBio™
QP10328-ec-250ug
250 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001014358.1
IL-31 / IL31 Protein
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SHTLPVRLLR PSDDVQKIVE ELQSLSKMLL KDVEEEKGVL VSQNYTLPCL SPDAQPPNNI HSPAIRAYLK TIRQLDNKSV IDEIIEHLDK LIFQDAPETN ISVPTDTHEC KRFILTISQQ FSECMDLALK SLTSGAQQAT T
Greater than 97% as determined by SDS-PAGE and HPLC.
386653
Research Use Only
IL31
The specific activity is determined by inducing STAT3 activation using human U-87 MG cells. 5 ng/mL of rHuIL-31 can effectively induce STAT3 activation.
Human
Untagged
0.2 um filtered PBS, pH7.4, lyophilized
Videos
SDS
Documents

Documents

Product Certifications