missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-31 / IL31 Protein

A DNA sequence encoding the Human IL-31 / IL31, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP10328-ec-2ug

 View more versions of this product

Product Code. 15963518

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001014358.1
IL-31 / IL31 Protein
Research Use Only
IL31
The specific activity is determined by inducing STAT3 activation using human U-87 MG cells. 5 ng/mL of rHuIL-31 can effectively induce STAT3 activation.
Human
Untagged
0.2 um filtered PBS, pH7.4, lyophilized
386653
2 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SHTLPVRLLR PSDDVQKIVE ELQSLSKMLL KDVEEEKGVL VSQNYTLPCL SPDAQPPNNI HSPAIRAYLK TIRQLDNKSV IDEIIEHLDK LIFQDAPETN ISVPTDTHEC KRFILTISQQ FSECMDLALK SLTSGAQQAT T
Greater than 97% as determined by SDS-PAGE and HPLC.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers