missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL16 / Interleukin-16 Protein
Recombinant Protein
Brand: enQuireBio™ QP5266-1mg
This item is not returnable.
View return policy
Specifications
NP_001165599.1 | |
IL16 / Interleukin-16 Protein | |
Research Use Only | |
IL16 | |
Recombinant Protein | |
E. coli | |
MPDLNSSTDS AASASAASDV SVESTAEATV CTVTLEKMSA GLGFSLEGGK GSLHGDKPLT INRIFKGAAS EQSETVQPGD EILQLGGTAM QGLTRFEAWN IIKALPDGPV TIVIRRKSLQ SKETTAAGDS | |
Greater than 95% as determined by SDS-PAGE. |
3603 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human IL16 / Interleukin-16 induces Primary human T cell chemotaxis. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human IL16 / Interleukin-16 Protein > 1mg
Spot an opportunity for improvement?Share a Content Correction