missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL16 / Interleukin-16 Protein

Recombinant Protein

53285.00 SEK - 87550.00 SEK

Specifications

Accession Number NP_001165599.1
Gene ID (Entrez) 3603
Name IL16 / Interleukin-16 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15915809
View Documents
enQuireBio™
QP5266-500ug
500 μg
53285.00 SEK
500µg
Please to purchase this item. Need a web account? Register with us today!
15905809
View Documents
enQuireBio™
QP5266-1mg
1 mg
87550.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001165599.1
IL16 / Interleukin-16 Protein
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Human IL16 / Interleukin-16 induces Primary human T cell chemotaxis.
Human
Untagged
10 mM sodium phosphate, pH 7.5
3603
Research Use Only
IL16
Recombinant Protein
E. coli
MPDLNSSTDS AASASAASDV SVESTAEATV CTVTLEKMSA GLGFSLEGGK GSLHGDKPLT INRIFKGAAS EQSETVQPGD EILQLGGTAM QGLTRFEAWN IIKALPDGPV TIVIRRKSLQ SKETTAAGDS
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human IL16 / Interleukin-16 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.