missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL1F10 / IL-38 Protein

A DNA sequence encoding the Homo sapiens (Human) IL1F10 / IL-38, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7890-ec-1mg

 View more versions of this product

Product Code. 16031421

  • 28125.00 SEK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_115945.4
IL1F10 / IL-38 Protein
Research Use Only
Il1f10
Recombinant Protein
E. coli
MCSLPMARYY IIKYADQKAL YTRDGQLLVG DPVADNCCAE KICILPNRGL ARTKVPIFLG IQGGSRCLAC VETEEGPSLQ LEDVNIEELY KGGEEATRFT FFQSSSGSAF RLEAAAWPGW FLCGPAEPQQ PVQLTKESEP SARTKFYFEQ SW
Greater than 90% as determined by SDS-PAGE.
84639
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.