missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1F10 / IL-38 Protein
A DNA sequence encoding the Homo sapiens (Human) IL1F10 / IL-38, was expressed in the hosts and tags indicated.
1580.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_115945.4 |
---|---|
Gene ID (Entrez) | 84639 |
Name | IL1F10 / IL-38 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16021421
|
enQuireBio™
QP7890-ec-10ug |
10 μg | N/A | ||||||
16061421
|
enQuireBio™
QP7890-ec-50ug |
50 μg | N/A | ||||||
16011421
|
enQuireBio™
QP7890-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16041421
|
enQuireBio™
QP7890-ec-200ug |
200 μg | N/A | ||||||
16051421
|
enQuireBio™
QP7890-ec-500ug |
500 μg | N/A | ||||||
16031421
|
enQuireBio™
QP7890-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_115945.4 | |
IL1F10 / IL-38 Protein | |
Not determined. | |
Full Length | |
Human | |
Tris-based buffer, 50% glycerol |
84639 | |
Research Use Only | |
Il1f10 | |
Recombinant Protein | |
MCSLPMARYY IIKYADQKAL YTRDGQLLVG DPVADNCCAE KICILPNRGL ARTKVPIFLG IQGGSRCLAC VETEEGPSLQ LEDVNIEELY KGGEEATRFT FFQSSSGSAF RLEAAAWPGW FLCGPAEPQQ PVQLTKESEP SARTKFYFEQ SW |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title