missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable ATP-dependent RNA helicase DDX58 Protein
A DNA sequence encoding the Homo sapiens (Human) Probable ATP-dependent RNA helicase DDX58, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5922-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_055129.2 | |
Probable ATP-dependent RNA helicase DDX58 Protein | |
Research Use Only | |
DDX58 | |
Recombinant Protein | |
E. coli | |
MTTEQRRSLQ AFQDYIRKTL DPTYILSYMA PWFREEEVQY IQAEKNNKGP MEAATLFLKF LLELQEEGWF RGFLDALDHA GYSGLYEAIE SWDFKKIEKL EEYRLLLKRL QPEFKTRIIP TDIISDLSEC LINQECEEIL QICSTKGMMA GAEKLVECLL RSDKENWPKT LKLALEKERN KFSELWIVEK GIKDVETEDL EDKMETSDIQ IFYQEDPECQ NLSENSCPPS EVSDTNLYSP FKPRNYQLEL ALPAMKGKNT IICAPTGCGK TFVSLLICEH HLKKFPQGQK GKVVFFANQI PVYEQQKSVF SKYFERHGYR VTGISGATAE NVPVEQIVEN NDIIILTPQI LVNNLKKGTI PSLSIFTLMI FDECHNTSKQ HPYNMIMFNY LDQKLGGSSG PLPQVIGLTA SVGVGDAKNT DEALDYICKL | |
Greater than 90% as determined by SDS-PAGE. |
23586 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction