missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Probable ATP-dependent RNA helicase DDX58 Protein
A DNA sequence encoding the Homo sapiens (Human) Probable ATP-dependent RNA helicase DDX58, was expressed in the hosts and tags indicated.
1640.00 SEK - 28125.00 SEK
Specifications
Accession Number | NP_055129.2 |
---|---|
Gene ID (Entrez) | 23586 |
Name | Probable ATP-dependent RNA helicase DDX58 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15968699
|
enQuireBio™
QP5922-ec-10ug |
10 μg | N/A | ||||||
15908709
|
enQuireBio™
QP5922-ec-50ug |
50 μg | N/A | ||||||
15958699
|
enQuireBio™
QP5922-ec-100ug |
100 μg |
6235.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15988699
|
enQuireBio™
QP5922-ec-200ug |
200 μg | N/A | ||||||
15998699
|
enQuireBio™
QP5922-ec-500ug |
500 μg | N/A | ||||||
15978699
|
enQuireBio™
QP5922-ec-1mg |
1 mg |
28125.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_055129.2 | |
Probable ATP-dependent RNA helicase DDX58 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
Tris-based buffer, 50% glycerol |
23586 | |
Research Use Only | |
DDX58 | |
Recombinant Protein | |
MTTEQRRSLQ AFQDYIRKTL DPTYILSYMA PWFREEEVQY IQAEKNNKGP MEAATLFLKF LLELQEEGWF RGFLDALDHA GYSGLYEAIE SWDFKKIEKL EEYRLLLKRL QPEFKTRIIP TDIISDLSEC LINQECEEIL QICSTKGMMA GAEKLVECLL RSDKENWPKT LKLALEKERN KFSELWIVEK GIKDVETEDL EDKMETSDIQ IFYQEDPECQ NLSENSCPPS EVSDTNLYSP FKPRNYQLEL ALPAMKGKNT IICAPTGCGK TFVSLLICEH HLKKFPQGQK GKVVFFANQI PVYEQQKSVF SKYFERHGYR VTGISGATAE NVPVEQIVEN NDIIILTPQI LVNNLKKGTI PSLSIFTLMI FDECHNTSKQ HPYNMIMFNY LDQKLGGSSG PLPQVIGLTA SVGVGDAKNT DEALDYICKL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title