missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protoporphyrinogen oxidase Protein

A DNA sequence encoding the Homo sapiens (Human) Protoporphyrinogen oxidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6521-ec-10ug

 View more versions of this product

Product Code. 16013890

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_000300.1
Protoporphyrinogen oxidase Protein
Research Use Only
PPOX
Recombinant Protein
E. coli
MGRTVVVLGG GISGLAASYH LSRAPCPPKV VLVESSERLG GWIRSVRGPN GAIFELGPRG IRPAGALGAR TLLLVSELGL DSEVLPVRGD HPAAQNRFLY VGGALHALPT GLRGLLRPSP PFSKPLFWAG LRELTKPRGK EPDETVHSFA QRRLGPEVAS LAMDSLCRGV FAGNSRELSI RSCFPSLFQA EQTHRSILLG LLLGAGRTPQ PDSALIRQAL AERWSQWSLR GGLEMLPQAL ETHLTSRGVS VLRGQPVCGL SLQAEGRWKV SLRDSSLEAD HVISAIPASV LSELLPAEAA PLARALSAIT AVSVAVVNLQ YQGAHLPVQG FGHLVPSSED PGVLGIVYDS VAFPEQDGSP PGLRVTVMLG GSWLQTLEAS GCVLSQELFQ QRAQEAAATQ LGLKEMPSHC LVHLHKNCIP QYTLGHWQKL ESARQFLTAH RLPLTLAGAS YEGVAVNDCI ESGRQAAVSV LGTEPNS
Greater than 90% as determined by SDS-PAGE.
5498
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers