missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protoporphyrinogen oxidase Protein

A DNA sequence encoding the Homo sapiens (Human) Protoporphyrinogen oxidase, was expressed in the hosts and tags indicated.

1640.00 SEK - 28125.00 SEK

Specifications

Accession Number NP_000300.1
Gene ID (Entrez) 5498
Name Protoporphyrinogen oxidase Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16013890
View Documents Promotion Details
enQuireBio™
QP6521-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16053890
View Documents Promotion Details
enQuireBio™
QP6521-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16003890
View Documents Promotion Details
enQuireBio™
QP6521-ec-100ug
100 μg
6235.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16033890
View Documents Promotion Details
enQuireBio™
QP6521-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16043890
View Documents Promotion Details
enQuireBio™
QP6521-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16023890
View Documents Promotion Details
enQuireBio™
QP6521-ec-1mg
1 mg
28125.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_000300.1
Protoporphyrinogen oxidase Protein
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
5498
Research Use Only
PPOX
Recombinant Protein
E. coli
MGRTVVVLGG GISGLAASYH LSRAPCPPKV VLVESSERLG GWIRSVRGPN GAIFELGPRG IRPAGALGAR TLLLVSELGL DSEVLPVRGD HPAAQNRFLY VGGALHALPT GLRGLLRPSP PFSKPLFWAG LRELTKPRGK EPDETVHSFA QRRLGPEVAS LAMDSLCRGV FAGNSRELSI RSCFPSLFQA EQTHRSILLG LLLGAGRTPQ PDSALIRQAL AERWSQWSLR GGLEMLPQAL ETHLTSRGVS VLRGQPVCGL SLQAEGRWKV SLRDSSLEAD HVISAIPASV LSELLPAEAA PLARALSAIT AVSVAVVNLQ YQGAHLPVQG FGHLVPSSED PGVLGIVYDS VAFPEQDGSP PGLRVTVMLG GSWLQTLEAS GCVLSQELFQ QRAQEAAATQ LGLKEMPSHC LVHLHKNCIP QYTLGHWQKL ESARQFLTAH RLPLTLAGAS YEGVAVNDCI ESGRQAAVSV LGTEPNS
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications