Learn More
Novus Biologicals Recombinant Human SOD1/Cu-Zn SOD Pre-formed Fibrils Protein
Brand: Novus Biologicals NBP3-14783-200UG
This item is not returnable.
View return policy
Specifications
Electron Microscopy, In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot | |
6647 | |
Store at -80C. Avoid freeze-thaw cycles. | |
SOD1 | |
>95%, by SDS-PAGE |
PB pH 7.4, 5 mM EDTA, 50 mM DTT | |
2 x 100 μg Vials | |
ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic | |
Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
SOD1/Cu-Zn SOD |