missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Human SOD1/Cu-Zn SOD Pre-formed Fibrils Protein
6110.00 SEK - 19930.00 SEK
Specifications
For Use With (Application) | Electron Microscopy, In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot |
---|---|
Formulation | PB pH 7.4, 5 mM EDTA, 50 mM DTT |
Gene ID (Entrez) | 6647 |
Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
Gene Alias | ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18321415
|
Novus Biologicals
NBP3-14783-100UG |
100 μg |
6110.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18310837
|
Novus Biologicals
NBP3-14783-200UG |
2 x 100 μg Vials |
9810.00 SEK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18391646
|
Novus Biologicals
NBP3-14783-500UG |
5 x 100 μg Vials |
19930.00 SEK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Electron Microscopy, In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot | |
6647 | |
ALS, ALS1, amyotrophic lateral sclerosis 1 (adult), Cu, Cu/Zn superoxide dismutase, EC 1.15.1.1, homodimer, hSod1, indophenoloxidase A, IPOA, SOD, SOD, soluble, superoxide dismutase [Cu-Zn], Superoxide dismutase 1, superoxide dismutase 1, soluble, Zn superoxide dismutase, EC 1.15.1.110superoxide dismutase, cystolic | |
Biologically Active Proteins, Cancer, Diabetes Research, Mitochondrial Fusion Proteins, Neuronal Cell Markers, Neuroscience, Tumor Suppressors | |
SOD1/Cu-Zn SOD |
PB pH 7.4, 5 mM EDTA, 50 mM DTT | |
Store at -80C. Avoid freeze-thaw cycles. | |
SOD1 | |
>95%, by SDS-PAGE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title