missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thioredoxin domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Thioredoxin domain-containing protein 5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7950-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_001139021.1 | |
Thioredoxin domain-containing protein 1 | |
Research Use Only | |
TXNDC5 | |
Recombinant Protein | |
E. coli | |
MEDAKVYVAK VDCTAHSDVC SAQGVRGYPT LKLFKPGQEA VKYQGPRDFQ TLENWMLQTL NEEPVTPEPE VEPPSAPELK QGLYELSASN FELHVAQGDH FIKFFAPWCG HCKALAPTWE QLALGLEHSE TVKIGKVDCT QHYELCSGNQ VRGYPTLLWF RDGKKVDQYK GKRDLESLRE YVESQLQRTE TGATETVTPS EAPVLAAEPE ADKGTVLALT ENNFDDTIAE GITFIKFYAP WCGHCKTLAP TWEELSKKEF PGLAGVKIAE VDCTAERNIC SKYSVRGYPT LLLFRGGKKV SEHSGGRDLD SLHRFVLSQA KDEL | |
Greater than 80% as determined by SDS-PAGE. |
81567 | |
200 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction