missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thioredoxin domain-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Thioredoxin domain-containing protein 5, was expressed in the hosts and tags indicated.

2430.00 SEK - 40175.00 SEK

Specifications

Accession Number NP_001139021.1
Gene ID (Entrez) 81567
Name Thioredoxin domain-containing protein 1
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16001801
View Documents
enQuireBio™
QP7950-ec-10ug
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16041801
View Documents
enQuireBio™
QP7950-ec-50ug
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16091791
View Documents
enQuireBio™
QP7950-ec-100ug
100 μg
9055.00 SEK
100µg
Please to purchase this item. Need a web account? Register with us today!
16021801
View Documents
enQuireBio™
QP7950-ec-200ug
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16031801
View Documents
enQuireBio™
QP7950-ec-500ug
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16011801
View Documents
enQuireBio™
QP7950-ec-1mg
1 mg
40175.00 SEK
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_001139021.1
Thioredoxin domain-containing protein 1
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
81567
Research Use Only
TXNDC5
Recombinant Protein
E. coli
MEDAKVYVAK VDCTAHSDVC SAQGVRGYPT LKLFKPGQEA VKYQGPRDFQ TLENWMLQTL NEEPVTPEPE VEPPSAPELK QGLYELSASN FELHVAQGDH FIKFFAPWCG HCKALAPTWE QLALGLEHSE TVKIGKVDCT QHYELCSGNQ VRGYPTLLWF RDGKKVDQYK GKRDLESLRE YVESQLQRTE TGATETVTPS EAPVLAAEPE ADKGTVLALT ENNFDDTIAE GITFIKFYAP WCGHCKTLAP TWEELSKKEF PGLAGVKIAE VDCTAERNIC SKYSVRGYPT LLLFRGGKKV SEHSGGRDLD SLHRFVLSQA KDEL
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.