Learn More
enQuireBio™ Recombinant Human Thioredoxin domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Thioredoxin domain-containing protein 5, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7950-ec-50ug
 View more versions of this product
Product Code. 16041801
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Specifications
NP_001139021.1
Thioredoxin domain-containing protein 1
Research Use Only
TXNDC5
Recombinant Protein
E. coli
MEDAKVYVAK VDCTAHSDVC SAQGVRGYPT LKLFKPGQEA VKYQGPRDFQ TLENWMLQTL NEEPVTPEPE VEPPSAPELK QGLYELSASN FELHVAQGDH FIKFFAPWCG HCKALAPTWE QLALGLEHSE TVKIGKVDCT QHYELCSGNQ VRGYPTLLWF RDGKKVDQYK GKRDLESLRE YVESQLQRTE TGATETVTPS EAPVLAAEPE ADKGTVLALT ENNFDDTIAE GITFIKFYAP WCGHCKTLAP TWEELSKKEF PGLAGVKIAE VDCTAERNIC SKYSVRGYPT LLLFRGGKKV SEHSGGRDLD SLHRFVLSQA KDEL
Greater than 80% as determined by SDS-PAGE.
81567
50 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol