missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat BD 4 Protein
A cDNA sequence encoding the BD 4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10534-1mg
This item is not returnable.
View return policy
Specifications
64389 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its antimicrobial activity against E. coli. The ED50 for this effect is typically 5-50microg/ml. | |
Rat | |
Untagged | |
BD-4 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
BD 4 Protein | |
Research Use Only | |
Defb4 | |
Recombinant Protein | |
E. coli | |
QSINNPITCL TKGGVCWGPC TGGFRQIGTC GLPRVRCCKK K | |
Greater than 97.0% as determined by SDS-PAGE. |