missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat BD 4 Protein
A cDNA sequence encoding the BD 4 was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 46290.00 SEK
Specifications
Gene ID (Entrez) | 64389 |
---|---|
Name | BD 4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | Defb4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15924168
|
enQuireBio™
QP10534-5ug |
5 μg |
1545.00 SEK
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15914168
|
enQuireBio™
QP10534-20ug |
20 μg |
2405.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15904168
|
enQuireBio™
QP10534-1mg |
1 mg |
46290.00 SEK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
64389 | |
Research Use Only | |
Defb4 | |
Recombinant Protein | |
E. coli | |
QSINNPITCL TKGGVCWGPC TGGFRQIGTC GLPRVRCCKK K | |
Greater than 97.0% as determined by SDS-PAGE. |
BD 4 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Measured by its antimicrobial activity against E. coli. The ED50 for this effect is typically 5-50microg/ml. | |
Rat | |
Untagged | |
BD-4 protein was lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |