Learn More
MP Biomedicals™ Red Screw Caps
Use with empty FastPrep™ tubes. Red Caps are screw scaps with an O-ring for use with 2 mL microfuge tubes. These caps are sold separately for those who wish to fill their own Lysing Matrix C tubes for prepare samples for homogenization
Brand: MP Biomedicals™ 115066005
Additional Details : Weight : 0.34000kg
Description
- Red Caps fit empty FastPrep™ tubes and are used in FastPrep systems.
- Add sample to empty tube, add Lysing Matrix C material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample.
- Red caps are also found in the Fast RNA™ Pro Red Kit for isolation of total RNA from yeast and fungi.
Specifications
Red | |
Store at Room Temperature (15°C to 30°C) | |
Screw Cap |
2 mL | |
1000 Pk. |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.