missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Red Screw Caps
Use with empty FastPrep™ tubes. Red Caps are screw scaps with an O-ring for use with 2 mL microfuge tubes. These caps are sold separately for those who wish to fill their own Lysing Matrix C tubes for prepare samples for homogenization
3535.10 SEK - 16795.70 SEK
Specifications
Color | Red |
---|---|
Volume (Metric) | 2 mL |
Storage Requirements | Store at Room Temperature (15°C to 30°C) |
Product Type | Screw Cap |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482340
|
MP Biomedicals™
115066005 |
1000 Pk. |
3535.10 SEK
Pack of 1000 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11492340
|
MP Biomedicals™
115066010 |
5000 Pk. |
16795.70 SEK
Pack of 5000 |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Red Caps fit empty FastPrep™ tubes and are used in FastPrep systems.
- Add sample to empty tube, add Lysing Matrix C material and lysing buffer, and apply cap. Process using FastPrep instrument or other bead-beater to lsye, grind or homogenize sample.
- Red caps are also found in the Fast RNA™ Pro Red Kit for isolation of total RNA from yeast and fungi.
Specifications
Red | |
2 mL | |
Store at Room Temperature (15°C to 30°C) | |
Screw Cap |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.