Learn More
Lenz Laborglasintrumente Reduction Adapters with Ground Joints
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01502445
This item is not returnable.
View return policy
Description
- Acc. to DIN 12257 and in addition to DIN standard
- Conical ground sockets and cones
- Made of DURAN™ tubing
Specifications
NS 45/40 | |
Borosilicate Glass (3.3) | |
Socket NS 24/29 |