Learn More
Lenz Laborglasintrumente Reduction Adapters with Ground Joints
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
108.12 SEK - 1118.30 SEK
Product Code | Brand | Standard Taper Joint No. | Cone Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Cone Size | Price | Quantity & Availability | ||||
10060201
|
Lenz Laborglasintrumente
01503419 |
Socket NS 34/35 | NS 19/26 |
378.42 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
10002933
|
Lenz Laborglasintrumente
01504571 |
Socket NS 45/40 | NS 71/51 |
1118.30 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
10707331
|
Lenz Laborglasintrumente
01503429 |
Socket NS 34/35 | NS 29/32 |
357.22 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
10070351
|
Lenz Laborglasintrumente
01502971 |
Socket NS 29/32 | NS 71/51 |
1075.90 SEK
Each |
Estimated Shipment: 27-09-2024
Log in to see stock. |
||||
10224051
|
Lenz Laborglasintrumente
01504560 |
Socket NS 45/40 | NS 60/46 |
764.26 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
10274571
|
Lenz Laborglasintrumente
01502445 |
Socket NS 24/29 | NS 45/40 |
333.90 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
10284331
|
Lenz Laborglasintrumente
01501934 |
Socket NS 19/26 | NS 34/35 |
258.64 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
10385451
|
Lenz Laborglasintrumente
01504534 |
Socket NS 45/40 | NS 34/35 |
465.34 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
10684192
|
Lenz Laborglasintrumente
01503424 |
Socket NS 34/35 | NS 24/29 |
425.06 SEK
Each |
Estimated Shipment: 01-10-2024
Log in to see stock. |
||||
10517701
|
Lenz Laborglasintrumente
01501429 |
Socket NS 14/23 | NS 29/32 |
108.12 SEK
Each |
In Stock
Log in to see stock. |
||||
Description
- Acc. to DIN 12257 and in addition to DIN standard
- Conical ground sockets and cones
- Made of DURAN™ tubing