Learn More
Fisherbrand™ Reinforced Silicone Tubing
Providing excellent crushing resistance and tear strength across a wide range of applications. Fisherbrand™ Reinforced Silicone Tubing is oderless and non-toxic.
Brand:  Fisherbrand™ 1174-3974
 View more versions of this product
Product Code. 11743974
  • 15300.00 SEK / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Autoclavable
  • Glossy in appearance
  • Translucent and flexible
  • FDA (food) compliant
  • Applications include pharmaceutical, high pressure and temperature work, biotechnology and aerospace
Specifications
Specifications
FDA, BFR and WRAS approved
3 mm
25 m
60 bar
Autoclavable
8.0 mm
2.5 mm