Learn More
Fisherbrand™ Reinforced Silicone Tubing
Providing excellent crushing resistance and tear strength across a wide range of applications. Fisherbrand™ Reinforced Silicone Tubing is oderless and non-toxic.
Brand:  Fisherbrand™ LRESI12519
 View more versions of this product
Product Code. 11793974
  • 33850.00 SEK / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Autoclavable
  • Glossy in appearance
  • Translucent and flexible
  • FDA (food) compliant
  • Applications include pharmaceutical, high pressure and temperature work, biotechnology and aerospace
Specifications
Specifications
FDA, BFR and WRAS approved
12.5 mm
25 m
30 bar
Autoclavable
18.9 mm
3.2 mm