missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11237-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the Rho family of GTPases. The encoded protein is localized to the outer mitochondrial membrane and plays a role in mitochondrial trafficking and fusion-fission dynamics.Specifications
RHOT2 | |
Polyclonal | |
Unconjugated | |
RHOT2 | |
ARHT2, C16orf39, hMiro 2, MIRO 2, miro2, Mitochondrial Rho GTPase 2, RASL, RHOT2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
214952, 89941 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8IXI1, Q8JZN7 | |
RHOT2 | |
RHOT2 Fusion Protein Ag1752 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |