missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOT2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RHOT2 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16898173
|
Proteintech
11237-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16888173
|
Proteintech
11237-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the Rho family of GTPases. The encoded protein is localized to the outer mitochondrial membrane and plays a role in mitochondrial trafficking and fusion-fission dynamics.Specifications
RHOT2 | |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8IXI1, Q8JZN7 | |
214952, 89941 | |
RHOT2 Fusion Protein Ag1752 | |
Primary | |
-20°C | |
RHOT2 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ARHT2, C16orf39, hMiro 2, MIRO 2, miro2, Mitochondrial Rho GTPase 2, RASL, RHOT2 | |
RHOT2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |