Learn More
Eurofins DiscoverX Ros Protein, Active
Active, N-terminal His6-tagged recombinant human Ros residues 1883-end, for use in Kinase Assays.
Brand: Eurofins DiscoverX 14-527M
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
Protein tyrosine kinase ROS is a role for morphogenic induction and proliferation of epithelial cells in the kidney, intestine and lung. ROS may provide a signal transduction pathway for epithelial-mesenchymal interactions.
GenBank M34353. The recombinant protein contains the conflict V2262A with respect toGenBank M34353. This is reported inGenBank NM_002944 and Z98880.
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
250 μg | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥60% |
53.4 kDa | |
Human | |
N-terminal 6His | |
Truncated | |
Ros, active |