missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eurofins DiscoverX Ros Protein, Active
Active, N-terminal His6-tagged recombinant human Ros residues 1883-end, for use in Kinase Assays.
Specifications
Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 53.4 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal 6His |
Description
Protein tyrosine kinase ROS is a role for morphogenic induction and proliferation of epithelial cells in the kidney, intestine and lung. ROS may provide a signal transduction pathway for epithelial-mesenchymal interactions.
GenBank M34353. The recombinant protein contains the conflict V2262A with respect toGenBank M34353. This is reported inGenBank NM_002944 and Z98880.
Specifications
mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal 6His | |
Truncated | |
Ros, active |
53.4 kDa | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥60% |